Sodium tricyanomethanide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Tricyanomethanide sodium salt

O0489-25G O0489-5G
TCO0489-25GEA 387 USD
TCO0489-25G TCO0489-5G
Sodium tricyanomethanide ≥98.0% (by titrimetric analysis)
Sodium tricyanomethanide
Formula: C₄N₃Na
MW: 113.05 g/mol
MDL Number: MFCD00174290
CAS Number: 36603-80-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR