4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene ≥98.0% (by GC)

Supplier: TCI America

B4102-1G B4102-5G
TCB4102-1GEA 513.7 USD
TCB4102-1G TCB4102-5G
4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene ≥98.0% (by GC)
4,4-Bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene
Formula: C₂₅H₃₈S₂
Storage Temperature: Refrigerator
MDL Number: MFCD28291964
CAS Number: 365547-20-2

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR