4-Ethynyl-4'-propylbiphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Ethynyl-4'-propyl-1,1'-biphenyl

E1231-1G E1231-5G
TCE1231-1GEA 109 USD
TCE1231-1G TCE1231-5G
4-Ethynyl-4'-propylbiphenyl ≥98.0% (by GC)
4-Ethynyl-4'-propylbiphenyl
Formula: C₁₇H₁₆
MW: 220.32 g/mol
MDL Number: MFCD18072535
CAS Number: 360768-57-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR