N,N'-Di(1-naphthyl)-N,N',9,9-tetraphenyl-9H-fluorene-2,7-diamine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: DPFL-NPB, N2,N7-Di(naphthalen-1-yl)-N2,N7,9,9-tetraphenyl-9H-fluorene-2,7-diamine, 9,9-Diphenyl-2,7-bis[N-(1-naphthyl)anilino]fluorene

D5443-1G D5443-200MG
TCD5443-1GEA 255 USD
TCD5443-1G TCD5443-200MG
N,N'-Di(1-naphthyl)-N,N',9,9-tetraphenyl-9H-fluorene-2,7-diamine ≥98.0% (by HPLC)
N,N'-Di(1-naphthyl)-N,N',9,9-tetraphenyl-9H-fluorene-2,7-diamine
Formula: C₅₇H₄₀N₂
MDL Number: MFCD11112145
CAS Number: 357645-40-0

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 237.0 to 241.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR