Melperone ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 4'-Fluoro-4-(4-methylpiperidino)butyrophenone

M2622-100MG M2622-25MG
TCM2622-100MGEA 338.96 USD
TCM2622-100MG TCM2622-25MG
Melperone ≥95.0% (by GC)
Melperone
Formula: C₁₆H₂₂FNO
MW: 263.36 g/mol
Boiling Pt: 125°C
Storage Temperature: Refrigerator
MDL Number: MFCD00867737
CAS Number: 3575-80-2
Merck Index: 14,05827

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR