9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 9,9-Bis(2-ethylhexyl)fluorene-2,7-diboronic acid bis(pinacol) ester

B4892-1G B4892-200MG
TCB4892-1GEA 579 USD
TCB4892-1G TCB4892-200MG
9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene ≥98.0% (by HPLC)
9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene
Formula: C₄₁H₆₄B₂O₄
MDL Number: MFCD16621099
CAS Number: 357219-41-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR