1-Methylimidazole bis(trifluoromethanesulfonyl)imide ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1-Methylimidazole Bis(trifluoromethanesulphonyl)imide, 1-Methylimidazolium bis(trifluoromethanesulphonyl)imide

M3210-25G M3210-5G
TCM3210-25GEA 339 USD
TCM3210-25G TCM3210-5G
1-Methylimidazole bis(trifluoromethanesulfonyl)imide ≥97.0% (by HPLC)
1-Methylimidazole bis(trifluoromethanesulfonyl)imide
Formula: C₆H₇F₆N₃O₄S₂
MDL Number: MFCD18251286
CAS Number: 353239-08-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 47.0 to 51.0 deg-C
Water max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR