N-(4'-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N-(4'-Bromo-[1,1'-biphenyl]-4-yl)-N-phenylnaphthalen-1-amine

B5049-1G B5049-200MG
TCB5049-1GEA 216.6 USD
TCB5049-1G TCB5049-200MG
N-(4'-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)
N-(4'-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine
Formula: C₂₈H₂₀BrN
MW: 450.38 g/mol
MDL Number: MFCD24387457
CAS Number: 352359-42-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR