Carbinoxamine Maleate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Carbinoxamine maleate salt
C3057-25G
C3057-5G
TCC3057-25GEA
220
USD
TCC3057-25G
TCC3057-5G
Carbinoxamine Maleate ≥98.0% (by HPLC, titration analysis)
Carbinoxamine Maleate
Formula:
C₁₆H₁₉ClN₂O·C₄H₄O₄ MW: 406.86 g/mol |
MDL Number:
MFCD00082461 CAS Number: 3505-38-2 EINECS: 222-498-0 UN: 2811 ADR: 6.1 III Merck Index: 14,01799 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 % (total of Ion-Pair) |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 121.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...