2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Ethynylboronic acid Pinacol ester

E1074-1G E1074-200MG
TCE1074-1GEA 241 USD
TCE1074-1G TCE1074-200MG
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0% (by GC)
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₈H₁₃BO₂
Boiling Pt: 140°C
Storage Temperature: Refrigerator
MDL Number: MFCD13182122
CAS Number: 347389-74-6
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR