(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97.0% (by GC)

Supplier: TCI America

Synonyms: (2S,5S)-5-Benzyl-2-(tert-butyl)-3-methylimidazolidin-4-one, (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone

B4138-1G B4138-200MG
76219-372EA 467 USD
76219-372 76219-374
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97.0% (by GC)
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone
Formula: C₁₅H₂₂N₂O
MW: 246.35 g/mol
MDL Number: MFCD03426982
CAS Number: 346440-54-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 98.0 to 102.0 deg-C
Specific rotation [a]20/D -58.0 to -63.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR