1-Methyl-3-pentylimidazolium bromide ≥98.0% (by HPLC)

Warning

Synonyms: 1-Methyl-3-pentyl-1H-imidazol-3-ium bromide

M3037-25G M3037-5G
76220-334 76220-336
1-Methyl-3-pentylimidazolium bromide ≥98.0% (by HPLC)
1-Methyl-3-pentylimidazolium bromide
Formula: C₉H₁₇BrN₂
MW: 233.15 g/mol
MDL Number: MFCD19382537
CAS Number: 343851-31-0

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR