N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D4152-1G
D4152-5G
TCD4152-1GEA
79
USD
TCD4152-1G
TCD4152-5G
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
N,N'-Di(4-pyridyl)-1,4,5,8-naphthalenetetracarboxdiimide
Formula:
C₂₄H₁₂N₄O₄ Melting Pt: 353°C |
MDL Number:
MFCD00326861 CAS Number: 34151-49-0 |
Specification Test Results
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...