O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: HOTU, N-(6-Cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)-N-methylmethanaminium hexafluorophosphate(V)

E1306-1G E1306-5G
TCE1306-1GEA 30 USD
TCE1306-1G TCE1306-5G
O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate
Formula: C₁₀H₁₇F₆N₄O₃P
Storage Temperature: Refrigerator
MDL Number: MFCD01862832
CAS Number: 333717-40-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR