Tiaprofenic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
T3175-100MG
T3175-25MG
TCT3175-100MGEA
425.8
USD
TCT3175-100MG
TCT3175-25MG
Tiaprofenic acid ≥98.0% (by GC, titration analysis)
Tiaprofenic acid
Formula:
C₁₄H₁₂O₃S |
MDL Number:
MFCD00866089 CAS Number: 33005-95-7 EINECS: 251-329-3 UN: 2811 ADR: 6.1,III Merck Index: 14,09422 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...