N,N-Dimethyl-4-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D5607-1G D5607-5G
TCD5607-1GEA 144 USD
TCD5607-1G TCD5607-5G
N,N-Dimethyl-4-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-4-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
MDL Number: MFCD22053163
CAS Number: 329-17-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR