Tris(1,10-phenanthroline)cobalt(II) bis(hexafluorophosphate) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Tris(1,10-phenanthroline)cobalt bis(hexafluorophosphate)

T3553-1G T3553-5G
TCT3553-1GEA 153 USD
TCT3553-1G TCT3553-5G
Tris(1,10-phenanthroline)cobalt(II) bis(hexafluorophosphate) ≥98.0% (by HPLC, titration analysis)
Tris(1,10-phenanthroline)cobalt(II) bis(hexafluorophosphate)
Formula: C₃₆H₂₄CoF₁₂N₆P₂
MW: 889.49 g/mol
MDL Number: MFCD30749310
CAS Number: 31876-74-1

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Chelometric Titration) min. 98.0 %
Solubility in Acetonitrile almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR