1,8-Bis(diphenylphosphinyl)naphthalene ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B5280-1G B5280-5G
76219-560EA 163 USD
76219-560 76219-562
1,8-Bis(diphenylphosphinyl)naphthalene ≥98.0% (by HPLC)
1,8-Bis(diphenylphosphinyl)naphthalene
Formula: C₃₄H₂₆O₂P₂
CAS Number: 316808-41-0

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR