2-(4-Bromophenyl)benzoxazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(4-Bromophenyl)benzo[d]oxazole

B5017-1G B5017-5G
76219-488EA 78 USD
76219-488 76219-490
2-(4-Bromophenyl)benzoxazole ≥98.0% (by GC)
2-(4-Bromophenyl)benzoxazole
Formula: C₁₃H₈BrNO
MW: 274.12 g/mol
MDL Number: MFCD00168898
CAS Number: 3164-13-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR