Fargesin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: rel-5-((1S,3aR,4R,6aR)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl)benzo[d][1,3]dioxole, rel-5-[(1R,3aS,4S,6aS)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-1,3-benzodioxole

F1188-10MG F1188-50MG
TCF1188-10MGEA 188 USD
TCF1188-10MG TCF1188-50MG
Fargesin ≥98.0% (by HPLC)
Fargesin
Formula: C₂₁H₂₂O₆
MW: 370.40 g/mol
MDL Number: MFCD07781423
CAS Number: 31008-19-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 132.0 to 136.0 deg-C
Specific rotation [a]20/D +113 to +125 deg (C=1, CH2CL2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR