3-Fluorothiophene ≥98.0% (by GC)

Supplier: TCI America
Danger

F0512-1G F0512-200MG
TCF0512-1GEA 830.6 USD
TCF0512-1G TCF0512-200MG
3-Fluorothiophene ≥98.0% (by GC)
3-Fluorothiophene
Formula: C₄H₃FS
MW: 102.13 g/mol
Boiling Pt: 85°C
Storage Temperature: Freezer
MDL Number: MFCD08275189
CAS Number: 3093-88-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR