Tributylmethylammonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3692-25G T3692-5G
TCT3692-25GEA 243 USD
TCT3692-25G TCT3692-5G
Tributylmethylammonium iodide ≥98.0% (by titrimetric analysis)
Tributylmethylammonium iodide
Formula: C₁₃H₃₀IN
MDL Number: MFCD00031623
CAS Number: 3085-79-8
EINECS: 221-402-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR