4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl ≥98.0% (by GC)

Supplier: TCI America

D5129-1G D5129-5G
TCD5129-1GEA 80 USD
TCD5129-1G TCD5129-5G
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl ≥98.0% (by GC)
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl
Formula: C₂₁H₁₃F₇O
MDL Number: MFCD22376741
CAS Number: 303186-19-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 67.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR