2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-((6-Chloro-2-methylpyrimidin-4-yl)amino)-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide, N-(2-Chloro-6-methylphenyl)-2-[(6-chloro-2-methyl-4-pyrimidinyl)amino]-5-thiazolecarboxamide

C3675-1G C3675-5G
TCC3675-1GEA 44 USD
TCC3675-1G TCC3675-5G
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by HPLC)
2-[(6-Chloro-2-methylpyrimidin-4-yl)amino]-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Formula: C₁₆H₁₃Cl₂N₅OS
MW: 394.27 g/mol
Melting Pt: 280°C
Storage Temperature: Refrigerator
MDL Number: MFCD11045075
CAS Number: 302964-08-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR