4-Benzyloxy-3-methoxyphenylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4800-1G B4800-5G
TCB4800-1GEA 161 USD
TCB4800-1G TCB4800-5G
4-Benzyloxy-3-methoxyphenylacetic acid ≥98.0% (by GC, titration analysis)
4-Benzyloxy-3-methoxyphenylacetic acid
Formula: C₁₆H₁₆O₄
MDL Number: MFCD00185562
CAS Number: 29973-91-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR