Isopropylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

I1041-1G I1041-5G
TCI1041-1GEA 34 USD
TCI1041-1G TCI1041-5G
Isopropylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)
Isopropylamine hydrobromide
Formula: C₃H₉N·HBr
MDL Number: MFCD22419143
CAS Number: 29552-58-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR