S-p-Tolyl p-Toluenesulfonothioate ≥98.0% (by HPLC)

Supplier: TCI America

T3173-1G T3173-5G
TCT3173-1GEA 160.69 USD
TCT3173-1G TCT3173-5G
S-p-Tolyl p-Toluenesulfonothioate ≥98.0% (by HPLC)
S-p-Tolyl p-Toluenesulfonothioate
Formula: C₁₄H₁₄O₂S₂
MDL Number: MFCD00092126
CAS Number: 2943-42-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR