4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine ≥95.0% (by GC)

Supplier: TCI America
Warning

T3597-1G T3597-5G
TCT3597-1GEA 105 USD
TCT3597-1G TCT3597-5G
4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine ≥95.0% (by GC)
4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine
Formula: C₉H₁₈F₃NO₂Si
MW: 257.33 g/mol
CAS Number: 289706-46-3

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR