2-Bromo-9,9-diethylfluorene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-9,9-diethyl-9H-fluorene

B5126-1G B5126-5G
TCB5126-1GEA 94 USD
TCB5126-1G TCB5126-5G
2-Bromo-9,9-diethylfluorene ≥98.0% (by GC)
2-Bromo-9,9-diethylfluorene
Formula: C₁₇H₁₇Br
MW: 301.23 g/mol
Boiling Pt: 163°C
MDL Number: MFCD09909863
CAS Number: 287493-15-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR