Cetraxate hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-[4-[[trans-4-(Aminomethyl)cyclohexyl]carbonyloxy]phenyl]propanoic acid hydrochloride

C3454-250MG C3454-50MG
TCC3454-250MGEA 461 USD
TCC3454-250MG TCC3454-50MG
Cetraxate hydrochloride ≥98.0% (by HPLC, titration analysis)
Cetraxate hydrochloride
Formula: C₁₇H₂₃NO₄·HCl
MW: 341.83 g/mol
Melting Pt: 240°C
MDL Number: MFCD01722166
CAS Number: 27724-96-5
Merck Index: 14,02024

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR