2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine ≥97.0% (by total nitrogen basis)
Supplier: TCI America
|
Warning
|
B4399-25G
B4399-5G
TCB4399-25GEA
359
USD
TCB4399-25G
TCB4399-5G
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine ≥97.0% (by total nitrogen basis)
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine
Formula:
C₃₃H₃₉N₃O₂ |
MDL Number:
MFCD09834146 CAS Number: 2725-22-6 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...