Disodium 3,3'-Dithiobis(1-propanesulfonate) ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D5159-100G D5159-25G
TCD5159-100GEA 89.45 USD
TCD5159-100G TCD5159-25G
Disodium 3,3'-Dithiobis(1-propanesulfonate) ≥97.0% (by titrimetric analysis)
Disodium 3,3'-Dithiobis(1-propanesulfonate)
Formula: C₆H₁₂Na₂O₆S₄
MDL Number: MFCD01104840
CAS Number: 27206-35-5
EINECS: 248-324-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR