9-(4-Ethynylphenyl)carbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

E1055-1G E1055-5G
TCE1055-1GEA 128 USD
TCE1055-1G TCE1055-5G
9-(4-Ethynylphenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
9-(4-Ethynylphenyl)carbazole
Formula: C₂₀H₁₃N
MW: 267.33 g/mol
MDL Number: MFCD12024288
CAS Number: 262861-81-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR