(8-Quinolinolato)lithium ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Lithium quinolin-8-olate, Liq

Q0100-1G Q0100-5G
TCQ0100-1GEA 81 USD
TCQ0100-1G TCQ0100-5G
(8-Quinolinolato)lithium ≥98.0% (by HPLC, titration analysis)
(8-Quinolinolato)lithium
Formula: C₉H₆LiNO
MW: 151.09 g/mol
MDL Number: MFCD00152778
CAS Number: 25387-93-3
EINECS: 246-932-3

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR