1-(2,6-Diisopropylphenyl)imidazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D4948-1G D4948-5G
76219-078EA 177 USD
76219-078 76219-080
1-(2,6-Diisopropylphenyl)imidazole ≥98.0% (by titrimetric analysis)
1-(2,6-Diisopropylphenyl)imidazole
Formula: C₁₅H₂₀N₂
MW: 228.34 g/mol
Boiling Pt: 105°C
MDL Number: MFCD13299721
CAS Number: 25364-47-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR