Dipentaerythritol Hexakis(3-mercaptopropionate) ≥93.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
DiPETMP, Dipentaerythritol hexa(3-mercaptopropionate)
D5212-25G
D5212-500G
TCD5212-25GEA
47
USD
TCD5212-25G
TCD5212-500G
Dipentaerythritol Hexakis(3-mercaptopropionate) ≥93.0% (by titrimetric analysis)
Dipentaerythritol Hexakis(3-mercaptopropionate)
Formula:
C₂₈H₄₆O₁₃S₆ MW: 783.02 g/mol |
MDL Number:
MFCD00128217 CAS Number: 25359-71-1 EINECS: 246-893-2 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Iodometric back titration) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...