4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: A-205804, 4-[(4-Methylphenyl)thio]thieno[2,3-c]pyridine-2-carboxamide, A 205804

T3498-100MG T3498-25MG
TCT3498-100MGEA 440 USD
TCT3498-100MG TCT3498-25MG
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide ≥95.0% (by HPLC)
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide
Formula: C₁₅H₁₂N₂OS₂
MW: 300.39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09038566
CAS Number: 251992-66-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 8.80 to 9.80 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR