2',3,4-Trifluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2',3,4-Trifluoro-4''-propyl-p-terphenyl

T3419-1G T3419-5G
76220-438EA 99 USD
76220-438 76220-440
2',3,4-Trifluoro-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
2',3,4-Trifluoro-4''-propyl-1,1':4',1''-terphenyl
Formula: C₂₁H₁₇F₃
MW: 326.36 g/mol
MDL Number: MFCD20488039
CAS Number: 248936-60-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 71.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR