9H-Pyrido[2,3-b]indole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: alpha-Carboline

P2310-1G P2310-200MG
TCP2310-1GEA 424 USD
TCP2310-1G TCP2310-200MG
9H-Pyrido[2,3-b]indole ≥98.0% (by GC, titration analysis)
9H-Pyrido[2,3-b]indole
Formula: C₁₁H₈N₂
MW: 168.20 g/mol
Boiling Pt: 364°C
MDL Number: MFCD00473709
CAS Number: 244-76-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 214.0 to 218.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR