1-Methyl-3-propylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: 1-Methyl-3-propylimidazolium Tetrafluoroborate

M3036-25G M3036-5G
TCM3036-25GEA 168 USD
TCM3036-25G TCM3036-5G
1-Methyl-3-propylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1-Methyl-3-propylimidazolium tetrafluoroborate
Formula: C₇H₁₃BF₄N₂
MW: 212.00 g/mol
Melting Pt: 17°C
MDL Number: MFCD14156016
CAS Number: 244193-48-4
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR