Solifenacin Succinate ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
(S)-(R)-Quinuclidin-3-yl 1-phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate succinate
S0944-100MG
S0944-25MG
TCS0944-100MGEA
154
USD
TCS0944-100MG
TCS0944-25MG
Solifenacin Succinate ≥98.0% (by HPLC)
Solifenacin Succinate
Formula:
C₂₃H₂₆N₂O₂·C₄H₆O₄ MW: 480.56 g/mol |
MDL Number:
MFCD09952287 CAS Number: 242478-38-2 Merck Index: 14,08712 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 % (total of Ion-Pair) |
Melting point | 145.0 to 149.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...