3,3-Bis(bromomethyl)oxetane ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 2,2-Bis(bromomethyl)-1,3-epoxypropane

B5641-1G B5641-5G
TCB5641-1GEA 114 USD
TCB5641-1G TCB5641-5G
3,3-Bis(bromomethyl)oxetane ≥95.0% (by GC)
3,3-Bis(bromomethyl)oxetane
Formula: C₅H₈Br₂O
MW: 243.93 g/mol
Boiling Pt: 125°C
Melting Pt: 25°C
MDL Number: MFCD00269694
CAS Number: 2402-83-7

Order Now

Specification Test Results

Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR