11H-Benzo[a]carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,2-Benzocarbazole

B4635-1G B4635-5G
76219-414EA 155 USD
76219-414 76219-416
11H-Benzo[a]carbazole ≥98.0% (by GC)
11H-Benzo[a]carbazole
Formula: C₁₆H₁₁N
MW: 217.27 g/mol
MDL Number: MFCD00215939
CAS Number: 239-01-0
EINECS: 205-945-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 231.0 to 235.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR