Phenolphthalein Diphosphate Pentasodium salt hydrate ≥95.0% (by HPLC)

Supplier: TCI America

P2293-1G P2293-5G
TCP2293-1GEA 242 USD
TCP2293-1G TCP2293-5G
Phenolphthalein Diphosphate Pentasodium salt hydrate ≥95.0% (by HPLC)
Phenolphthalein Diphosphate Pentasodium salt hydrate
Formula: C₂₀H₁₃Na₅O₁₁P₂·xH₂O
MW: 606.21 g/mol
Storage Temperature: Refrigerator
CAS Number: 2382-56-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 95.0 area%
Water max. 30.0 %
Ethanol (NMR) max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR