4-(3-Pyridyl)phenylboronic acid hydrochloride (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

P2392-1G P2392-5G
TCP2392-1GEA 232 USD
TCP2392-1G TCP2392-5G
4-(3-Pyridyl)phenylboronic acid hydrochloride (contains varying amounts of Anhydride)
4-(3-Pyridyl)phenylboronic acid hydrochloride
Formula: C₁₁H₁₀BNO₂·HCl
CAS Number: 2377369-59-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) 97.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR