6-Chloro-4-quinolinol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Chloroquinolin-4-ol, 6-Chloro-4-hydroxyquinoline

C3312-1G C3312-5G
TCC3312-1GEA 38 USD
TCC3312-1G TCC3312-5G
6-Chloro-4-quinolinol ≥98.0% (by GC)
6-Chloro-4-quinolinol
Formula: C₉H₆ClNO
MW: 179.60 g/mol
Melting Pt: 269°C
MDL Number: MFCD00024011
CAS Number: 23432-43-1

Order Now

Specification Test Results

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR