Ethylene Glycol bis(3-mercaptopropionate) ≥98.0% (by titrimetric analysis), purified

Supplier: TCI America
Warning

Synonyms: Ethylene bis(3-mercaptopropionate), Glycol bis(3-mercaptopropionate)

E1329-100G E1329-25G
TCE1329-100GEA 474.96 USD
TCE1329-100G TCE1329-25G
Ethylene Glycol bis(3-mercaptopropionate) ≥98.0% (by titrimetric analysis), purified
Ethylene Glycol bis(3-mercaptopropionate)
Formula: C₈H₁₄O₄S₂
MW: 238.32 g/mol
Flash Pt: 255°C
MDL Number: MFCD00053520
CAS Number: 22504-50-3
EINECS: 245-044-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Iodometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR