5-Adamantyl-IAA ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 2-[5-(Adamantan-1-yl)-1H-indol-3-yl]acetic acid, [5-(Tricyclo[3.3.1.1~3,7~]decan-1-yl)-1H-indol-3-yl]acetic acid

A3390-10MG A3390-50MG
TCA3390-10MGEA 171 USD
TCA3390-10MG TCA3390-50MG
5-Adamantyl-IAA ≥98.0% (by HPLC)
5-Adamantyl-IAA
Formula: C₂₀H₂₃NO₂
MW: 309.41 g/mol
Storage Temperature: Refrigerator
CAS Number: 2244426-40-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR