4-(tert-Butoxycarbonylamino)cyclohexanol ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: (4-Hydroxycyclohexyl)carbamic acid tert-Butyl ester, tert-Butyl (4-Hydroxycyclohexyl)carbamate, 4-(Boc-amino)cyclohexanol

B4714-1G B4714-5G
TCB4714-1GEA 34.5 USD
TCB4714-1G TCB4714-5G
4-(tert-Butoxycarbonylamino)cyclohexanol ≥98.0% (by GC, total nitrogen)
4-(tert-Butoxycarbonylamino)cyclohexanol
Formula: C₁₁H₂₁NO₃
MDL Number: MFCD06658349
CAS Number: 224309-64-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR