Bicyclo[1.1.1]pentan-1-amine hydrochloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5479-1G B5479-200MG
TCB5479-1GEA 318 USD
TCB5479-1G TCB5479-200MG
Bicyclo[1.1.1]pentan-1-amine hydrochloride ≥98.0% (by GC, titration analysis)
Bicyclo[1.1.1]pentan-1-amine hydrochloride
Formula: C₅H₉N·HCl
MW: 119.59 g/mol
MDL Number: MFCD19204065
CAS Number: 22287-35-0

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR